missing translation for 'onlineSavingsMsg'
Learn More

Azelaic Acid,, Sigma-Aldrich™

Product Code. 15820761
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15820761 1 kg 1kg
15810761 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15820761 Supplier Sigma Aldrich Supplier No. 8201161000
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Technical grade for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-99-9
Molecular Formula HOOC(CH2)7COOH
Molecular Weight (g/mol) 188.22

Specifications

Melting Point 107°C
Boiling Point 237°C/20 hPa
Density 1.029 g/cm3 at 20°C
Quantity 1 kg
MDL Number MFCD00004432
Synonym Azelaic acid,, Nonanedioic acid, Azelaic acid,
Chemical Name or Material Azelaic Acid,
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.