Learn More
AZD8797, MedChemExpress
Product Code. 16488527
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16488527
Brand: Medchem Express HY13848
MedChemExpress AZD8797 (KAND567) is an allosteric non-competitive and orally active antagonist of the human CX3CR1 receptor; antagonizes CX3CR1 and CXCR2 with Kis of 3.9 and 2800 nM, respectively.
AZD8797 (KAND567) is an allosteric non-competitive and orally active antagonist of the human CX3CR1 receptor
Specifications
Chemical Name or Material AZD8797
Physical Form Solid
CAS 911715-90-7
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H25N5OS2
For Use With (Application) COVID-19-immunoregulation
Synonym KAND567
Solubility Information DMSO : ≥ 150 mg/mL (371.69 mM)
SMILES CC(C)C[C@@H](NC1=NC(S[C@@H](C)C2=CC=CC=C2)=NC3=C1SC(N)=N3)CO
Molecular Weight (g/mol) 403.56
Formula Weight 403.56
Percent Purity 98.65%
Grade Research