missing translation for 'onlineSavingsMsg'
Learn More

AZD8329, MedChemExpress

Product Code. p-200097483
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487436 100 mg 100mg
16463497 50 mg 50mg
16417478 10 mg 10mg
16409957 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16487436 Supplier Medchem Express Supplier No. HY18173
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AZD8329 is a potent 11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor with an IC50 of 9 nM for human 11β-HSD1, displays excellent selectivity versus 11β-HSD2, 17β-HSD1 and 17β-HSD3.

AZD8329 is a potent 11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor with an IC50 of 9 nM for human 11β-HSD1, displays excellent selectivity versus 11β-HSD2, 17β-HSD1 and 17β-HSD3.

Specifications

Chemical Name or Material AZD8329
Physical Form Solid
CAS 1048668-70-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C25H31N3O3
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information DMSO : 62.5 mg/mL (148.27 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(N2N=CC(C(NC3C(C4)CC5CC4CC3C5)=O)=C2C(C)(C)C)C=C1
Molecular Weight (g/mol) 421.53
Formula Weight 421.53
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.