Learn More
AZD7507, MedChemExpress
Product Code. 16497617
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497617 5 mg 5mg
16484868 10 mg 10mg
16483266 25 mg 25mg
16425656 100 mg 100mg
16421127 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16497617 Supplier Medchem Express Supplier No. HY117244
 Restricted Item
Estimated Shipment: 10-11-2026
MedChemExpress AZD7507 is a potent and orally active CSF-1R inhibitor, with antitumor activity.
AZD7507 is a potent and orally active CSF-1R inhibitor, with antitumor activity.
Specifications
Chemical Name or Material AZD7507
CAS 1041852-85-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H27FN6O3
Quantity 5 mg
Solubility Information DMSO : 130 mg/mL (286.03 mM; Need ultrasonic)
SMILES O=C(N)C1=NN=C2C=C(C(N3CCN(CC3)CCO)=CC2=C1NC4=CC=C(C=C4F)C)OC
Molecular Weight (g/mol) 454.5
Formula Weight 454.5
Percent Purity 99.14%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease