missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZD-9291 Mesylate, TRC
Specifications
Specifications
| Chemical Name or Material | Osimertinib Mesylate |
| CAS | 1421373-66-1 |
| Molecular Formula | C28 H33 N7 O2 . C H4 O3 S |
| Quantity | 100 mg |
| Synonym | 2-Propenamide, N-[2-[[2-(dimethylamino)ethyl]methylamino]-4-methoxy-5-[[4-(1-methyl-1H-indol-3-yl)-2-pyrimidinyl]amino]phenyl]-, methanesulfonate (1:1), AZD 9291 mesylate, Osimertinib mesylate, Osimertinib mesilate |
| InChI Formula | InChI=1S/C28H33N7O2.CH4O3S/c1-7-27(36)30-22-16-23(26(37-6)17-25(22)34(4)15-14-33(2)3)32-28-29-13-12-21(31-28)20-18-35(5)24-11-9-8-10-19(20)24;1-5(2,3)4/h7-13,16-18H,1,14-15H2,2-6H3,(H,30,36)(H,29,31,32);1H3,(H,2,3,4) |
| SMILES | COc1cc(N(C)CCN(C)C)c(NC(=O)C=C)cc1Nc2nccc(n2)c3cn(C)c4ccccc34.CS(=O)(=O)O |
| IUPAC Name | N-[2-[2-(dimethylamino)ethyl-methylamino]-4-methoxy-5-[[4-(1-methylindol-3-yl)pyrimidin-2-yl]amino]phenyl]prop-2-enamide;methanesulfonic acid |
| Molecular Weight (g/mol) | 595.71 |
| Formula Weight | 595.2577 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?