missing translation for 'onlineSavingsMsg'
Learn More

Azaperone, TRC

Product Code. 30007917
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007917 500 mg 500mg
18002450 10 mg 10mg
18060450 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007917 Supplier Toronto Research Chemicals Supplier No. TRCA802200500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1649-18-9
Molecular Formula C19 H22 F N3 O
Molecular Weight (g/mol) 327.4
Synonym Butyrophenone, 4'-fluoro-4-[4-(2-pyridyl)-1-piperazinyl]- (6CI,8CI), 1-(4-Fluorophenyl)-4-[4-(2-pyridinyl)-1-piperazinyl]-1-butanone, 4'-Fluoro-4-[4-(2-pyridyl)-1-piperazinyl]butyrophenone, Azaperone, Fluoperidol, NSC 170976, R-1929, Stresnil, Suicalm
IUPAC Name 1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one
SMILES Fc1ccc(cc1)C(=O)CCCN2CCN(CC2)c3ccccn3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H22FN3O/c20-17-8-6-16(7-9-17)18(24)4-3-11-22-12-14-23(15-13-22)19-5-1-2-10-21-19/h1-2,5-10H,3-4,11-15H2
Formula Weight 327.1747
Percent Purity >95%
Chemical Name or Material Azaperone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.