missing translation for 'onlineSavingsMsg'
Learn More

Azaperone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30311046
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311046 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311046 Supplier Dr. Ehrenstorfer Supplier No. DREA10340510AL100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azaperone
CAS 1649-18-9
Analyte or Component Names Azaperone
Shipping Condition Room Temperature
Molecular Formula C19 H22 F N3 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Azaperone, 1-(4-Fluorophenyl)-4-[4-(pyridin-2-yl)piperazin-1-yl]butan-1-one, 1-Butanone, 1-(4-fluorophenyl)-4-[4-(2-pyridinyl)-1-piperazinyl]-, Butyrophenone, 4'-fluoro-4-[4-(2-pyridyl)-1-piperazinyl]- (6CI,8CI), 4'-Fluoro-4-[4-(2-pyridyl)-1-piperazinyl]butyrophenone, Azaperone, Fluoperidol, NSC 170976, R-1929, Stresnil, Suicalm
InChI Formula InChI=1S/C19H22FN3O/c20-17-8-6-16(7-9-17)18(24)4-3-11-22-12-14-23(15-13-22)19-5-1-2-10-21-19/h1-2,5-10H,3-4,11-15H2
SMILES Fc1ccc(cc1)C(=O)CCCN2CCN(CC2)c3ccccn3
IUPAC Name 1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 327.4
Quantity 1 mL
Formula Weight 327.1747 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.