Learn More
Axygen™ Presterilized Conical Centrifuge Tubes
GREENER_CHOICE
Product Code. 12569067 Shop All Axygen Scientific Products
Change view
Click to view available options
Capacity (Metric):
15 mL
50 mL
Packaging:
25 Tubes and Caps/Rack, 20 Racks/Cs.
25 Tubes and Caps/Unit, 20 Units/Cs.
50 Tubes and Caps/Unit, 20 Units/Cs.
Bag
Unit Size:
Pack of 25
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Packaging unitSize
12569067 15 mL 50 Tubes and Caps/Unit, 20 Units/Cs. Pack of 500
12519107 50 mL 25 Tubes and Caps/Unit, 20 Units/Cs. Pack of 500
11344205 50 mL 25 Tubes and Caps/Unit, 20 Units/Cs. Pack of 25
12549067 15 mL Bag Pack of 500
11314175 15 mL Bag Pack of 25
12539107 50 mL 25 Tubes and Caps/Rack, 20 Racks/Cs. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12569067 Supplier Axygen™ Supplier No. SCT15MLRS
Add to basket
Add to basket
Graduated tubes with conical bottom
  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Screw Cap
Autoclavable Not Autoclavable
Capacity (Metric) 15 mL
Color Clear
Material Polypropylene
For Use With (Application) For sample identification
Includes Screw cap
Packaging 50 Tubes and Caps/Unit, 20 Units/Cs.
Sterility Sterile
Bottom Shape Conical
Max. RCF 12,000 x g