missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Clear Polypropylene Gel Loading Tip

Product Code. 10505540 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
0.5 to 200 μL
Unit Size:
Pack of 400
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10505540 0.5 to 200 μL Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10505540 Supplier Axygen™ Supplier No. TGL200FT37RS
Add to basket
Add to basket
This item is not returnable. View return policy

Designed for removing bands from agarose gel

Axygen™ Gel Loading Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brand of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. Available in either flat and round versions, with different outer diameters for different gel thickness.


  • Designed for removing bands from agarose gel
  • One handed operation
  • Push button gel release
  • Ejector button tip release
  • Avoid cross contamination
  • Tips fit standard 1000 μL pipettors
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • OD Value: 0.37mm
  • Sterile
  • TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 0.5 to 200 μL
    Sterility Sterile
    Color Clear
    Clean Claims DNase, RNase Free
    Format Rack
    Tip Style Gel Loading
    For Use With (Equipment) Universal Fit
    Material Polypropylene
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.