Learn More
Axygen™ Caps for PCR Tubes
Product Code. 12607385 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Assorted
Blue
Clear
Green
MaxyClear
MaxyClear, w/writing surface
Orange
Red
Violet
Yellow
Closure Type:
Domed Cap
Flat Cap
Product Type:
8-Strip Domed PCR Tube Cap
8-Strip Flat PCR Tube Cap
8-Strip Flat PCR Tube Cap with Writing Surface
PCR Caps
Unit Size:
Pack of 100
Pack of 125
Pack of 1250
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Closure Type Product Type Unit Size
12607385 Clear Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
10240900 Blue Domed Cap 8-Strip Domed PCR Tube Cap Pack of 1250
12547987 Blue Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12537987 Assorted Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12557987 Green Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12567987 Orange Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12587987 Violet Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12577987 Red Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12597987 Yellow Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12609535 MaxyClear, w/writing surface Flat Cap 8-Strip Flat PCR Tube Cap with Writing Surface Pack of 125
11320155 MaxyClear Flat Cap 8-Strip Flat PCR Tube Cap Pack of 125
15409073 Clear Domed Cap PCR Caps Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 12607385 Supplier Axygen™ Supplier No. PCR02CPC
  • ONLINE EXCLUSIVE PRICE
    30.30 EUR
    27.05 EUR / Pack of 125
    Save 3.25 EUR
    10% Off
Estimated Shipment: 15-12-2026
8-Strip domed or flat strip caps for Axygen PCR tubes
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Clear
Color Clear
Volume (Metric) 0.2 mL
Quantity 125/Pk.
Closure Material Polypropylene
For Use With (Equipment) 8-strips PCR tubes and 96-well PCR microplates
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Format Strip
Product Type 8-Strip Domed PCR Tube Cap