missing translation for 'onlineSavingsMsg'
Learn More

Avobenzone, TRC

Product Code. 30137133
Change view
Click to view available options
Quantity:
25 mg
1 g
10 g
Unit Size:
10g
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137133 10 g 10g
30138776 1 g 1g
17978354 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137133 Supplier Toronto Research Chemicals Supplier No. TRCA79469010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70356-09-1
Molecular Formula C20 H22 O3
Molecular Weight (g/mol) 310.39
Synonym 1-[4-(1,1-Dimethylethyl)phenyl]-3-(4-methoxyphenyl)-1,3-propanedione, 1-(4-Methoxyphenyl)-3-(4-tert-butylphenyl)propane-1,3-dione, 1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)propan-1,3-dione, 1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione, 1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione, 1-(4'-tert-Butylphenyl)-3-(4'-methoxyphenyl)propane-1,3-dione, 4-(1,1-Dimethylethyl)-4'-methoxydibenzoylmethane, 4-Methoxy-4'-tert-butyldibenzoylmethane, 4-tert-Butyl-4'-methoxydibenzoylmethane, Avobenzone, Butylmethoxydibenzoylmethane, Escalol 517, Eusolex 9020, NeoHeliopan 357, Parsol 1789, Parsol A, Parsol RTM 1789, Photoplex, Solarom BMBM, Uvinul BMBM
IUPAC Name 1-(4-tert-butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione
SMILES COc1ccc(cc1)C(=O)CC(=O)c2ccc(cc2)C(C)(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C20H22O3/c1-20(2,3)16-9-5-14(6-10-16)18(21)13-19(22)15-7-11-17(23-4)12-8-15/h5-12H,13H2,1-4H3
Formula Weight 310.1569
Percent Purity >95%
Chemical Name or Material Avobenzone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.