Learn More
Avanafil, MedChemExpress
Product Code. 16483507
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483507 50 mg 50mg
16477446 100 mg 100mg
16421896 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16483507 Supplier Medchem Express Supplier No. HY18252
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Avanafil (TA-1790) is a potent and selective phosphodiesterase-5 (PDE-5) inhibitor with IC50 values of 5.2 nM, 630 nM, 5700 nM, 6200 nM, 12000 nM, 27000 nM, 51000 nM and 53000 nM for PDE-5, PDE-6, PDE-4, PDE-10, PDE-8, PDE-7, PDE-2 and PDE-1, respectively.
Avanafil activates NO/cGMP/PKG signaling-pathway to decrease loss in BMD, bone atrophy, and oxidative stress. Avanafil inhibits cyclic guanosine monophosphate (cGMP) hydrolysis and thus increases cGMP levels. Avanafil can be used for the research of erectile dysfunction and osteoporosis.
Specifications
Chemical Name or Material Avanafil
CAS 330784-47-9
Color White
Purity Grade Notes Research
Molecular Formula C23H26ClN7O3
Quantity 50 mg
Synonym TA1790
Solubility Information DMSO : 50 mg/mL (103.32 mM; Need ultrasonic)
SMILES COC1=C(Cl)C=C(CNC2=C(C(NCC3=NC=CC=N3)=O)C=NC(N4[C@H](CO)CCC4)=N2)C=C1
Molecular Weight (g/mol) 483.95
Formula Weight 483.95
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation