missing translation for 'onlineSavingsMsg'
Learn More

AV-412, MedChemExpress

Product Code. p-200100846
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
50 mg
missing translation for 'unitSize':
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
16473848 10 mg 10mg
16426577 5 mg 5mg
16400227 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16473848 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY10346
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AV-412 (MP412) is an EGFR inhibitor with IC50s of 0.75, 0.5, 0.79, 2.3, 19 nM for EGFR, EGFRL858R, EGFRT790M, EGFRL858R/T790M and ErbB2, respectively.

AV-412 (MP412) is an EGFR inhibitor with IC50s of 0.75, 0.5, 0.79, 2.3, 19 nM for EGFR, EGFRL858R, EGFRT790M, EGFRL858R/T790M and ErbB2, respectively.

Specifications

Chemical Name or Material AV-412
CAS 451493-31-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C41H44ClFN6O7S2
Quantity 10 mg
Synonym MP412
Solubility Information DMSO : ≥ 28 mg/mL (32.89 mM)
SMILES O=S(C1=CC=C(C)C=C1)(O)=O.C=CC(NC2=CC3=C(NC4=CC=C(F)C(Cl)=C4)N=CN=C3C=C2C#CC(N5CCN(C)CC5)(C)C)=O.O=S(C6=CC=C(C)C=C6)(O)=O
Molecular Weight (g/mol) 851.41
Formula Weight 851.41
Percent Purity 98.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.