missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Autotaxin modulator 1, MedChemExpress
Description
Autotaxin modulator 1 is an autotaxin (ATX) enzyme inhibitor, extracted from patent WO 2014018881 A1, Compound Example 12b. Autotaxin modulator 1 is expected to be useful for researching demyelination due to injury or disease, as well as for researching proliferative disorders such as cancer.
Specifications
Specifications
| Chemical Name or Material | Autotaxin modulator 1 |
| CAS | 1548743-69-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C28H31F6NO3 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 100 mg/mL (183.98 mM; Need ultrasonic) |
| SMILES | O=C(C1CC(N2[C@@H](C3=CC=C4C=CC(O[C@H]5CC[C@@H](C(F)(F)F)CC5)=C(C(F)(F)F)C4=C3)C)CCC2C1)O |
| Molecular Weight (g/mol) | 543.54 |
| Formula Weight | 543.54 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?