Learn More
Velp Scientifica™ Autosampler Disc 2
Product Code. 15329584 Shop All Velp Products
Change view
Click to view available options
Type:
Autosampler Disc 2
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15329584 Autosampler Disc 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15329584 Supplier Velp Scientifica™ Supplier No. A00000199
  • ONLINE EXCLUSIVE PRICE
    326.65 EUR
    308.00 EUR / Pack of 1
    Save 18.65 EUR
    5% Off
Estimated Shipment: 20-10-2026
Velp Scientifica™ Autosampler Disc 2 for use with Dumas Nitrogen Analyzer NDA 701 and 702.
  • For use with NDA 701 and 702
Specifications
Type Autosampler Disc 2
For Use With Dumas Nitrogen Analyzer
Weight (Metric) 0.54 kg