Learn More
Applied Biosystems™ AutoLys M TubeLifter
Alle ansehen Applied Biosystems Produkte
missing translation for 'orderingAttributeHoverText'
Quantity:
1 each
missing translation for 'unitSize'
Each
Description
Once centrifugation is complete the rack of tubes is returned to the TubeLifter and the inner tube of each AutoLys M Tube is separated from the outer tube. The outer tube contains the lysate that is processed further using the MagMAX FFPE DNA/RNA Ultra Kit to recover the nucleic acids.
Specifications
Specifications
| For Use With (Equipment) | AutoLys TubeLifter System |
| Quantity | 1 each |
| Shipping Condition | Room Temperature |