Learn More
Abeba™ Autoclavable Clogs 9620
Product Code. 15673258
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Unit Size:
1 pair
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15673258 36 1 pair
15683258 37 1 pair
15693258 38 1 pair
15603268 39 1 pair
15613268 40 1 pair
15623268 41 1 pair
15633268 42 1 pair
15643268 43 1 pair
15653268 44 1 pair
15663268 45 1 pair
15673268 46 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15673258 Supplier Abeba™ Supplier No. 9620/35/36
  • ONLINE EXCLUSIVE PRICE
    50.25 EUR
    46.34 EUR / 1 pairs
    Save 3.91 EUR
    7% Off
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Antistatic and autoclavable at 121°C. High slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Green
  • Antistatic
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRC
Color Green
Size 36
Style 9620