Learn More
Abeba™ Autoclavable Clogs 39620
Product Code. 15621258
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Unit Size:
1 pair
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15621258 36 1 pair
15631258 37 1 pair
15641258 38 1 pair
15651258 39 1 pair
15661258 40 1 pair
15671258 41 1 pair
15681258 42 1 pair
15691258 43 1 pair
15601268 44 1 pair
15611268 45 1 pair
15621268 46 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15621258 Supplier Abeba™ Supplier No. 39620/35/36
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Antistatic and autoclavable at 135°C for 5 minutes. High slip resistance. Washing at 60°C.
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Green
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRC
Color Green
Size 36
Sole Features Slip-resistant
Style 39620