Learn More
3M™ Aura™ Particulate Respirator, Series 9400
Product Code. 12932505
Change view
Click to view available options
Filter Type:
FFP2
FFP3
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Type unitSize
12932505 FFP2 Pack of 10
12942505 FFP3 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12932505 Supplier 3M™ Supplier No. 7000088835
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
  • Provide reliable and comfortable protection from a range of airborne disposable hazards
  • Minimize contamination risks to food or pharmaceutical production processes
  • Made without staples or small detachable parts
  • Cool Flow™ exhalation valve option provides effective removal of heat and moisture build up for a more comfortable wear
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149:2001 + A1:2009
Color Blue
Filter Type FFP2
With Exhalation Valve Yes
Filter Efficiency FFP2
For Use With (Application) Exposure to airborne duest particles and non-volatile liquid particles
Includes Cool Flow™ exhalation valve
Material Headstrap Thermoplastic Elastomer
Nose Piece Material Aluminum
Resistant To Airborne dust and non-volatile liquid particles
Sterility Non-sterile
FDA Cleared Surgical Mask No