Learn More
Attune™ Wash Solution
Product Code. 17403822
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17403822 1 L 1 L
15360445 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17403822 Supplier Invitrogen Supplier No. J24974
 Restricted Item
In Stock
Add to basket
Add to basket
Attune™ Wash Solution is an optimized cleaning buffer that is used in the Attune™ Acoustic Focusing Cytometer to maintain cleanliness of the fluidics lines, minimizing contamination or carryover.
Maintains System Cleanliness
Attune™ Wash Solution has been formulated to remove cellular debris, prevent protein building up, and minimize dyes sticking to the fluidics lines of the system. It is recommended for use during the routine maintenance and automated shutdown procedures of the Attune™ Acoustic Focusing Cytometer.
Convenient
The ready-to-use 1X wash solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The wash solution is stable for 30 days after the bottle is opened.
High Quality
The Attune™ Wash Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
  • Attune™ Wash Solution is an optimized cleaning buffer that is used in the Attune™ Acoustic Focusing Cytometer
  • Helps maintain cleanliness of the fluidics lines to minimize contamination or carryover
  • Formulated to remove cellular debris, prevent protein building up, and minimize dyes sticking to the fluidics lines of the system
  • It is recommended for use during the routine maintenance and automated shutdown procedures of the Attune™ Acoustic Focusing Cytometer
  • The ready-to-use 1X wash solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system
  • Solution is stable for 30 days after the bottle is opened
  • The Attune™ Wash Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality
  • Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
For Use With (Equipment) Attune™ Acoustic Focusing Cytometer
Product Line Attune
Quantity 1 L
Product Type Wash Solution