Learn More
Invitrogen™ Attune™ NxT Waste BTL Tubing
Click to view available options
Quantity:
Each
Unit Size:
Each
Description
- This is replacement tubing for the waste bottle of the Attune NxT Acoustic Focusing Cytometer.
- The tubing can be replaced easily without replacing the entire bottle assembly if the tubing connector to the instrument is broken.
Specifications
Specifications
| Product Line | FPL |
| Quantity | Each |
| Format | Tubing |
| Shipping Condition | Room Temperature |
| For Use With (Equipment) | Attune NxT |
| Product Type | Tubing |