Learn More
Invitrogen™ Attune™ NxT Blocker Bar Conversion Kit
Description
- The Attune NxT Blocker Bar Conversion Kit provides a 1.5 mm blocking bar for use on the Attune NxT Acoustic Focusing Cytometer, which is delivered with a 0.5 mm blocking bar
- The standard 0.5 mm blocking bar accommodates cells or particles from 0.2–10 microns in size
- The larger blocking bar included in this kit is recommended for analysis of cells that are primarily larger than 10 microns
- A larger blocking bar blocks more forward scatter light and allows the detection of larger cells
- An experienced Thermo Fisher Scientific engineer can install the larger blocking bar on any Attune NxT cytometer, simplifying the instrument settings optimization
Specifications
Specifications
| Product Line | Attune |
| Content And Storage | • 1 blocker bar • 1 blocker bar holder |
| Quantity | 1 kit |
| Shipping Condition | Room Temperature |
| For Use With (Equipment) | Attune NxT Acoustic Focusing Cytometer |
| Product Type | Blocker Bar Conversion Kit |