Learn More
Applied Biosystems™ Attune™ Autosampler Waste Bottle
Description
- This is a replacement bottle
- For waste for the Attune™ Auto Sampler.
Specifications
Specifications
| Product Line | Attune |
| Content And Storage | Store at room temperature |
| Quantity | 1 each |
| Shipping Condition | Room Temperature |
| For Use With (Equipment) | Attune Acoustic Focusing Cytometer |
| Product Type | Autosampler Waste Bottle |