missing translation for 'onlineSavingsMsg'
Learn More

Atrazine, TRC

Product Code. 30133190
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133190 500 mg 500mg
30132523 100 mg 100mg
17939249 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133190 Supplier Toronto Research Chemicals Supplier No. TRCA794600500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1912-24-9
Molecular Formula C8 H14 Cl N5
Molecular Weight (g/mol) 215.68
Synonym 1,3,5-Triazine-2,4-diamine, 6-chloro-N-ethyl-N'-(1-methylethyl)- (9CI), s-Triazine, 2-chloro-4-(ethylamino)-6-(isopropylamino)- (8CI), 6-Chloro-N2-ethyl-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine, 1-Chloro-3-ethylamino-5-isopropylamino-2,4,6-triazine, 2-Chloro-4-(ethylamino)-6-(2-propylamino)-s-triazine, 2-Chloro-4-(ethylamino)-6-(isopropylamino)-s-triazine, 2-Chloro-4-(ethylamino)-6-(isopropylamino)triazine, 2-Chloro-4-ethylamineisopropylamine-s-triazine, 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine, 2-Ethylamino-4-isopropylamino-6-chloro-1,3,5-triazine, 2-Ethylamino-4-isopropylamino-6-chloro-s-triazine, 6-Chloro-4-(ethylamino)-2-(isopropylamino)-s-triazine, 6-Chloro-N-Ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diamine, A 361, AAtrex, AAtrex 4L, AAtrex Nine-O, ATZ, Aatrax, Aatrex 4LC, Aatrex L, Akticon, Aktikon, Aktikon PK, Aktinit A, Aktinit PK, Argezin, Atanor 50 SC, Atradex, Atragranz, Atranex, Atranex 80WP, Atraphyt, Atrasan, Atrataf, Atrazerba FL, Atrazin, Atrazine, Azoprim, Azoprim 50, CET, Chromozin, Cyazin, Farmozine, G 30027, Gesamprim, Gesaprim, Gesaprim 50, Gesaprim 500, Gesaprim L, Herbatoxol, Hungazin, Hungazin PK, NSC 163046, Nortox 500SC, Nu-Trazine, Oleogesaprim, Oleogesaprim 200, Primatol A, Primoleo, Radazin, Radazin T, Triazine A 1294, Wonuk, Zealin L, Zeapos, Zeazin, Zeazine, Zeopos
IUPAC Name 6-chloro-4-N-ethyl-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine
SMILES CCNc1nc(Cl)nc(NC(C)C)n1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)
Formula Weight 215.0938
Chemical Name or Material Atrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.