Learn More
Atrazine 200 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30319188
Change view
Click to view available options
Quantity:
4 x 1 mL
Unit Size:
4mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319188 4 x 1 mL 4mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30319188 Supplier Dr. Ehrenstorfer Supplier No. DREGS09010036ME
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Atrazine
CAS 1912-24-9
Analyte or Component Names Atrazine
Shipping Condition +20°C
Molecular Formula C8 H14 Cl N5
Concentration 200 μg/mL
Certifications/Compliance ISO 17034
Synonym 1,3,5-Triazine-2,4-diamine, 6-chloro-N2-ethyl-N4-(1-methylethyl)-, 1,3,5-Triazine-2,4-diamine, 6-chloro-N-ethyl-N'-(1-methylethyl)- (9CI), s-Triazine, 2-chloro-4-(ethylamino)-6-(isopropylamino)- (8CI), 6-Chloro-N2-ethyl-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine, 1-Chloro-3-ethylamino-5-isopropylamino-2,4,6-triazine, 2-Chloro-4-(ethylamino)-6-(2-propylamino)-s-triazine, 2-Chloro-4-(ethylamino)-6-(isopropylamino)-s-triazine, 2-Chloro-4-(ethylamino)-6-(isopropylamino)triazine, 2-Chloro-4-ethylamineisopropylamine-s-triazine, 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine, 2-Ethylamino-4-isopropylamino-6-chloro-1,3,5-triazine, 2-Ethylamino-4-isopropylamino-6-chloro-s-triazine, 6-Chloro-4-(ethylamino)-2-(isopropylamino)-s-triazine, 6-Chloro-N-Ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diamine, A 361, AAtrex, AAtrex 4L, AAtrex Nine-O, ATZ, Aatrax, Aatrex 4LC, Aatrex L, Akticon, Aktikon, Aktikon PK, Aktinit A, Aktinit PK, Argezin, Atradex, Atragranz, Atranex, Atranex 80WP, Atraphyt, Atrasan, Atrataf, Atrazerba FL, Atrazin, Atrazine, Azoprim, Azoprim 50, CET, Chromozin, Cyazin, Farmozine, G 30027, Gesamprim, Gesaprim, Gesaprim 50, Gesaprim 500, Gesaprim L, Herbatoxol, Hungazin, Hungazin PK, NSC 163046, Nortox 500SC, Nu-Trazine, Oleogesaprim, Oleogesaprim 200, Primatol A, Primoleo, Radazin, Radazin T, Triazine A 1294, Wonuk, Zealin L, Zeapos, Zeazin, Zeazine, Zeopos
InChI Formula InChI=1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)
SMILES CCNc1nc(Cl)nc(NC(C)C)n1
IUPAC Name 6-chloro-4-N-ethyl-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine
Solvent or Matrix Methanol
Molecular Weight (g/mol) 215.68
Quantity 4 x 1 mL
Formula Weight 215.0938 g/mol