missing translation for 'onlineSavingsMsg'
Learn More

Atractylenolide III, MedChemExpress

Product Code. p-200101932
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498888 10 mg 10mg
16464787 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498888 Supplier Medchem Express Supplier No. HYN0203
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Atractylenolide III is a major component of Atractylodes rhizome can induce apoptosis of the lung carcinoma cells.

Atractylenolide III is a major component of Atractylodes rhizome can induce apoptosis of the lung carcinoma cells.

Specifications

Chemical Name or Material Atractylenolide III
CAS 73030-71-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H20O3
Quantity 10 mg
Synonym ICodonolactone 8β-Hydroxyasterolide
Solubility Information DMSO : 50 mg/mL (201.35 mM; Need ultrasonic)
SMILES O=C1C(C)=C(C[C@@]23[H])[C@@](C[C@@]3(C)CCCC2=C)(O)O1
Molecular Weight (g/mol) 248.32
Formula Weight 248.32
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.