missing translation for 'onlineSavingsMsg'
Learn More

Atractylenolide I, MedChemExpress

Product Code. p-200096931
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491328 5 mg 5mg
16478888 10 mg 10mg
16444787 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491328 Supplier Medchem Express Supplier No. HYN0201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Atractylenolide I is a sesquiterpene derived from the rhizome of Atractylodes macrocephala, possesses diverse bioactivities, such as neuroprotective, anti-allergic, anti-inflammatory and anticancer properties. Atractylenolide I reduces protein levels of phosphorylated JAK2 and STAT3 in A375 cells, and acts as a TLR4-antagonizing agent.

Atractylenolide I is a sesquiterpene derived from the rhizome of Atractylodes macrocephala, possesses diverse bioactivities, such as neuroprotective, anti-allergic, anti-inflammatory and anticancer properties. Atractylenolide I reduces protein levels of phosphorylated JAK2 and STAT3 in A375 cells, and acts as a TLR4-antagonizing agent.

Specifications

Chemical Name or Material Atractylenolide I
CAS 73069-13-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H18O2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (434.22 mM; Need ultrasonic)
SMILES O=C1C(C)=C(C[C@@]23[H])C(O1)=C[C@@]3(C)CCCC2=C
Molecular Weight (g/mol) 230.3
Formula Weight 230.3
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.