missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atractylenolide I, MedChemExpress
Description
Atractylenolide I is a sesquiterpene derived from the rhizome of Atractylodes macrocephala, possesses diverse bioactivities, such as neuroprotective, anti-allergic, anti-inflammatory and anticancer properties. Atractylenolide I reduces protein levels of phosphorylated JAK2 and STAT3 in A375 cells, and acts as a TLR4-antagonizing agent.
Specifications
Specifications
| Chemical Name or Material | Atractylenolide I |
| CAS | 73069-13-3 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C15H18O2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (434.22 mM; Need ultrasonic) |
| SMILES | O=C1C(C)=C(C[C@@]23[H])C(O1)=C[C@@]3(C)CCCC2=C |
| Molecular Weight (g/mol) | 230.3 |
| Formula Weight | 230.3 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?