missing translation for 'onlineSavingsMsg'
Learn More

ATN-161 trifluoroacetate salt, MedChemExpress

Product Code. 16498417
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498417 5 mg 5mg
16481897 50 mg 50mg
16476216 100 mg 100mg
16475738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498417 Supplier Medchem Express Supplier No. HY13535A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ATN-161 trifluoroacetate salt is a novel integrin α5β1 antagonist, which inhibits angiogenesis and growth of liver metastases in a murine model.

ATN-161 trifluoroacetate salt is a novel integrin α5β1 antagonist, which inhibits angiogenesis and growth of liver metastases in a murine model.

Specifications

Chemical Name or Material ATN-161 trifluoroacetate salt
CAS 904763-27-5
Color White
Purity Grade Notes Research
Molecular Formula C25H36F3N9O10S
Quantity 5 mg
Synonym ATN-161 TFA salt
Solubility Information H2O : 5 mg/mL (7.03 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 1 mg/mL (1.41 mM; Need ultrasonic)
SMILES FC(F)(C(O)=O)F.O=C(N)C[C@@H](C(N)=O)NC([C@H](CS)NC([C@H](CO)NC([C@H](CC1=CNC=N1)NC([C@H]2N(C(C)=O)CCC2)=O)=O)=O)=O
Molecular Weight (g/mol) 711.67
Formula Weight 711.67
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.