Learn More
Biochrom Atlantis 2 Microplate Washer for 96 Well Plates with 2 Liquid Lines
Description
- Configurable to many different well formats for maximum flexibility
- Optional 16-way manifold for washing 384-well plates
- Detachable plate carrier for easy cleaning
- Low noise, pressure-free aspiration and dispensing pumps
- Auto rinse function prevents liquid path clogging
- Reduces aerosols due to cover and pumping technology
- Optional liquid level sensors
Specifications
Specifications
| Description | Atlantis 2 Microplate Washer for 96 Well Plates with 2 Liquid Lines |
| Dispense Volume | 50 to 2000 μL |
| Plate Type | 96-well Plate, 384-well Plate |
| Residual Volume | ±1 μL/well |
| Weight (Metric) | 6 kg |
| Display | 2 x 16 character display with backlight |
| For Use With (Application) | ELISA |
| Increments | 50 μL |
| Dimensions (L x W x H) | 46 x 21 x 21 cm |
| Weight (English) | 14 lb. |
| Show More |
For research use only