Learn More
Applied Biosystems™ ATC Full-skirted Adaptor
Product Code. 30182280 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 adaptor
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30182280 1 adaptor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30182280 Supplier Applied Biosystems™ Supplier No. A33045
 Restricted Item
Estimated Shipment: 11-03-2027
This full-skirted adaptor comes factory-installed on the 96-well Automated Thermal Cycler (ATC).
This full-skirted adaptor comes factory-installed on the 96-well Automated Thermal Cycler (ATC). It ensures that uniform pressure is applied by the heated lid onto the plate, and also ensures that the plate is ejected smoothly without sticking. This adaptor is only for use with 96-well fully skirted plates.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Automated Thermal Cycler (ATC)
Product Type ATC Full-skirted Plate Adaptor
For Use With (Application) End-point PCR
Quantity 1 adaptor