missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AT7519, MedChemExpress
Description
AT7519 (AT7519M) as a potent inhibitor of CDKs, with IC50s of 210, 47, 100, 13, 170, and <10 nM for CDK1, CDK2, CDK4 to CDK6, and CDK9, respectively.
Specifications
Specifications
| Chemical Name or Material | AT7519 |
| CAS | 844442-38-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H17Cl2N5O2 |
| Quantity | 50 mg |
| Synonym | AT7519M |
| Solubility Information | DMSO : ≥ 50 mg/mL (130.81 mM) |
| SMILES | O=C(NC1=CNN=C1C(NC2CCNCC2)=O)C3=C(Cl)C=CC=C3Cl |
| Molecular Weight (g/mol) | 382.24 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?