Learn More
AT-1002 TFA, MedChemExpress
Description
AT-1002 TFA, a 6-mer synthetic peptide, is a tight junction regulator and absorption enhancer.
Specifications
Specifications
| Chemical Name or Material | AT-1002 TFA |
| Physical Form | Solid |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C34H54F3N9O9S |
| For Use With (Application) | Metabolism-protein/nucleotide metabolism |
| Solubility Information | DMSO : 33.33 mg/mL (40.55 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.22 mM; Need ultrasonic) |
| SMILES | N[C@@H](CC1=CC=CC=C1)C(N[C@@H](CS)C(N[C@@H]([C@H](CC)C)C(NCC(N[C@@H](CCCNC(N)=N)C(N[C@@H](CC(C)C)C(O)=O)=O)=O)=O)=O)=O.OC(C(F)(F)F)=O |
| Molecular Weight (g/mol) | 821.91 |
| Show More |