missing translation for 'onlineSavingsMsg'
Learn More

AT-1002 TFA, MedChemExpress

Product Code. 16401097
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16401097 50 mg 50mg
16487577 5 mg 5mg
16445626 100 mg 100mg
16404838 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16401097 Supplier Medchem Express Supplier No. HY114426A50mg
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AT-1002 TFA, a 6-mer synthetic peptide, is a tight junction regulator and absorption enhancer.

AT-1002 TFA, a 6-mer synthetic peptide, is a tight junction regulator and absorption enhancer.

Specifications

Chemical Name or Material AT-1002 TFA
Physical Form Solid
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C34H54F3N9O9S
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information DMSO : 33.33 mg/mL (40.55 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.22 mM; Need ultrasonic)
SMILES N[C@@H](CC1=CC=CC=C1)C(N[C@@H](CS)C(N[C@@H]([C@H](CC)C)C(NCC(N[C@@H](CCCNC(N)=N)C(N[C@@H](CC(C)C)C(O)=O)=O)=O)=O)=O)=O.OC(C(F)(F)F)=O
Molecular Weight (g/mol) 821.91
Formula Weight 821.91
Percent Purity 98.82%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.