Learn More
Grant Instruments™ Replacement Hydrophobic Filter
Product Code. 13446468
Change view
Click to view available options
For Use With (Equipment):
FTA-1 Aspirator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13446468 FTA-1 Aspirator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13446468 Supplier Grant Instruments™ Supplier No. FA1
  • ONLINE EXCLUSIVE PRICE
    32.10 EUR
    30.21 EUR / Pack of 1
    Save 1.89 EUR
    5% Off
View all promotions
Estimated Shipment: 13-01-2027
Prevents contaminants from entering the pump.
  • For use with Grant FTA-1 Aspirator
  • 0.027 micron in-line filter prevents contaminants from entering the pump
  • Direction of flow indicated on filter body for correct installation
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) FTA-1 Aspirator