missing translation for 'onlineSavingsMsg'
Learn More

Asenapine maleate, MedChemExpress

Product Code. 16441016
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441016 200 mg 200mg
16465286 100 mg 100mg
16451596 500 mg 500mg
16410717 50 mg 50mg
16404398 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16441016 Supplier Medchem Express Supplier No. HY11100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Asenapine maleate is a 5-HT (1A, 1B, 2A, 2B, 2C, 5A, 6, 7) and D2 antagonist with Ki values of 0.03-4.0 nM, 1.3nM, respectively, and an antipsychotic.

Asenapine maleate is a 5-HT (1A, 1B, 2A, 2B, 2C, 5A, 6, 7) and D2 antagonist with Ki values of 0.03-4.0 nM, 1.3nM, respectively, and an antipsychotic.

Specifications

Chemical Name or Material Asenapine maleate
CAS 85650-56-2
Color White
Purity Grade Notes Research
Molecular Formula C21H20ClNO5
Quantity 200 mg
Synonym Org 5222 maleate
Solubility Information DMSO : 25 mg/mL (62.21 mM; Need ultrasonic) ∣H2O : 6.25 mg/mL (15.55 mM; Need ultrasonic and warming)
SMILES CN1C[C@]2([H])C3=CC=CC=C3OC4=CC=C(Cl)C=C4[C@]2(C1)[H].O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 401.84
Formula Weight 401.84
Percent Purity 95.07%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.