Learn More
Artesunate, MedChemExpress
Product Code. 16479026
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479026 100 mg 100mg
16434787 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16479026 Supplier Medchem Express Supplier No. HYN0193
 Restricted Item
Estimated Shipment: 10-11-2026
MedChemExpress Artesunate is an inhibitor of both STAT-3 and exported protein 1 (EXP1).
Artesunate is an inhibitor of both STAT-3 and exported protein 1 (EXP1).
Specifications
Chemical Name or Material Artesunate
CAS 88495-63-0
Color White
Purity Grade Notes Research
Molecular Formula C19H28O8
Quantity 100 mg
Solubility Information DMSO : 83.33 mg/mL (216.77 mM; Need ultrasonic) ∣7.5% sodium bicarbonate : 20 mg/mL (52.03 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O[C@H]1[C@H](C)[C@]2([H])CC[C@@H](C)[C@]3([H])CC[C@@](O4)(C)OO[C@]32[C@]4([H])O1)CCC(O)=O
Molecular Weight (g/mol) 384.42
Formula Weight 384.42
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease