missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo-lll, free acid, 80.85%, MP Biomedicals™

Product Code. 11480912 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480912 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11480912 Supplier MP Biomedicals Supplier No. 0219505905
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 1668-00-4
Molecular Formula C22H18As2N4O14S2
Synonym 2,7-bis(2-arsonophenylazo)-1, 8-dihydroxy-3, 6-naphthalene disulfonic acid, 2, 7-bis(2-arsonophenylazo)-1, 8-dihydroxy-3
Color Brown
Quantity 5 g
InChI Key TVMZRHVOFZTNET-RIRMOVKSSA-N
Physical Form Powder
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4[As](=O)(O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)[As](=O)(O)O
IUPAC Name (3Z,6E)-3,6-bis[(2-arsonophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
Molecular Weight (g/mol) 776.363
PubChem CID 9810878
Formula Weight 776.04
Percent Purity 80.85%
Chemical Name or Material Arsenazo-lll
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.