missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo III, Supelco™

Product Code. 15873891
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15873891 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15873891 Supplier Supelco Supplier No. 1101070005
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Metal indicator

TRUSTED_SUSTAINABILITY

Specifications

CAS 1668-00-4
MDL Number MFCD00036695
Synonym Arsenazo III, 2,7-Bis(2-arsonophenylazo)-1,8-dihydroxynaphthaline-3,6-disulfonic acid, Arsenazo III
Quantity 5 g
Molecular Weight (g/mol) 776.37
Grade Research
Chemical Name or Material Arsenazo III
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.