Learn More
Sartorius arium™ mini plus Pretreatment Cartridge
Product Code. 15525153 Shop All Sartorius Products
Change view
Click to view available options
Height (English):
10.2 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (English) Unit Size
15525153 10.2 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15525153 Supplier Sartorius Supplier No. H2OCPR
  • ONLINE EXCLUSIVE PRICE
    474.00 EUR
    373.00 EUR / Pack of 1
    Save 101.00 EUR
    21% Off
 Restricted Item
Estimated Shipment: 11-12-2026
For use with arium mini plus Ultrapure Water System
  • Protection for treatment of feed water
  • Fast and effective adsorption of impurities through high-grade activated
  • Efficient catalyst for removing oxidation agents such as chlorine
  • Efficient reverse osmosis membranes, optimized water consumption
  • Low-energy membranes for ecological and economical operation
TRUSTED_SUSTAINABILITY
Specifications
Description Sartorius arium™ mini plus Ultrapure Water System Accessory, Pre-treatment Cartridge
Capacity 7.7 lb.
Feedwater Source Tap
Material Polypropylene
Purification Method Activated Carbon
Resin Type Activated Carbon
Filter Type Catalytic activated carbon, Spherical
For Use With (Equipment) arium mini plus Ultrapure Water System
Type Pretreatment Cartridge
Width (English) 7 in.
Width (Metric) 18 cm
Height (English) 10.2 in.
Height (Metric) 26 cm
Depth (English) 4.3 in.
Depth (Metric) 11 cm
Weight (English) 7.7 lb.
Weight (Metric) 3.5 kg