missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ʟ-Arginine Monohydrochloride, Millipore™
Specifications
Specifications
| Melting Point | >235°C |
| MDL Number | MFCD00064550 |
| Synonym | α-Amino-δ-guanidino valerianic acid hydrochloride, alpha-Amino-delta-guanidino valerianic acid hydrochloride, Arg, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride |
| Density | 1.42 g/cm3 at 20°C |
| Molecular Weight (g/mol) | 210.66 |
| Quantity | 10 kg |
| Grade | Research |
| Chemical Name or Material | ʟ-Arginine monohydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?