missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Arg-Gly-Asp-Ser

Product Code. 16441307
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441307 Supplier MedChemExpress Supplier No. HY12290
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Arg-Gly-Asp-Ser is an integrin binding sequence that inhibits integrin receptor function. Arg-Gly-Asp-Ser directly and specifically bind pro-caspase-8, pro-caspase-9 and pro-caspase-3, while it does not bind pro-caspase-1.

Specifications

Purity 98.0%
CAS 91037-65-9
Content And Storage Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
For Use With (Application) COVID-19-immunoregulation
Molecular Weight (g/mol) 433.42 g/mol
Product Type Peptides
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 55 mg/mL (126.90 mM) ∣H2O : ≥ 25 mg/mL (57.68 mM)
Synonym RGDS peptide∣ Fibronectin tetrapeptide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.