missing translation for 'onlineSavingsMsg'
Learn More

ARFGAP1 Antibody, Novus Biologicals™

Product Code. 18603026 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18603026 25 μL 25µL
18180649 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18603026 Supplier Novus Biologicals Supplier No. NBP23878725ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARFGAP1 Polyclonal specifically detects ARFGAP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARFGAP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q8N6T3
Gene Alias ADP-ribosylation factor 1 GTPase activating protein, ADP-ribosylation factor 1 GTPase-activating protein, ADP-ribosylation factor GTPase activating protein 1, ADP-ribosylation factor GTPase-activating protein 1, ARF GAP 1, ARF1 GAP, ARF1-directed GTPase-activating protein, ARF1GAP, bA261N11.3, FLJ10767, GAP protein, HRIHFB2281, MGC39924
Gene Symbols ARFGAP1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: TLPSMVHRVSGQPQSVTASSDKAFEDWLNDDLGSYQGAQGNRYVGFGNTPPPQKKEDDFLNNAMSSLYSGWSSFTTGASRFASAAKEGATKFGSQASQKASEL
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55738
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.