Learn More
Aramchol, TRC
Specifications
Specifications
| Chemical Name or Material | Aramchol |
| CAS | 246529-22-6 |
| Molecular Formula | C44H79NO5 |
| Quantity | 25 mg |
| Synonym | (3β,5β,7α,12α)-7,12-Dihydroxy-3-[(1-oxoeicosyl)amino]-cholan-24-oic Acid, (3β)-Arachidylamido-7α,12α-dihydroxy-5β-cholan-24-oic Acid, C20-FABAC |
| InChI Formula | InChI=1S/C44H79NO5/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-40(48)45-34-27-28-43(3)33(29-34)30-38(46)42-36-25-24-35(32(2)23-26-41(49)50)44(36,4)39(47)31-37(42)43/h32-39,42,46-47H,5-31H2,1-4H3,(H,45,48)(H,49,50)/t32-,33+,34+,35-,36+,37+,38-,39+,42+,43+,44-/m1/s1 |
| SMILES | C[C@H](CCC(O)=O)[C@@]1([H])CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@@H](NC(CCCCCCCCCCCCCCCCCCC)=O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@@]21C |
| IUPAC Name | (4R)-4-[(3S,5S,7R,8R,9S,10S,12S,13R,14S,17R)-7,12-dihydroxy-3-(icosanoylamino)-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid |
| Molecular Weight (g/mol) | 702.1 |
| Formula Weight | 701.5958 |