missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Occurs in liver, brain, glandular organs, and depot fats of animals, in small amounts in human depot fats, and is a constituent of animal phosphatides.
- Arachidonic acid (AA) is an unsaturated ω6 fatty acid constituent of the phospholipids of cell membranes.
- Phospholipase A2 releases AA from the membrane phospholipids in response to inflammation.
- AA is subsequently metabolized to prostaglandins and thromboxanes by at least two cyclooxygenase (COX) isoforms, to leukotrienes and lipoxins by lipoxygenases, and to epoxyeicosatrienoic acids via cytochrome p450-catalyzed metabolism.
- AA and its metabolites play important roles in a variety of biological processes, including signal transduction, smooth muscle contraction, chemotaxis, cell proliferation and differentiation, and apoptosis.
- AA has been demonstrated to bind to the a subunit of G protein and inhibit the activity of Ras GTPase-activating proteins (GAPs).
- Cellular uptake of AA is energy dependent and involves protein-facilitated transport across the plasma membrane.
- Arachidonic acid stimulates adhesion of MDA-MB-435 human metastatic cancer cells to extracellular matrix molecules (collagen IV and vitronectin).
- Arachidonic acid plays a key role in cellular regulation and is controlled through multiple interconnected pathways.
- Changes in arachidonic acid levels correspond to the synthesis of eicosanoids resulting from receptor-stimulated lipid hydrolysis.
- It activates protein kinase C.
- Arachidonic acid serves as a precursor to numerous eicosanoids as well as other bioactive molecules.
Specifications
Specifications
| Color | Colorless |
| Density | 0.922g/mL |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Synonym | arachidonic acid, arachidonate, immunocytophyte, all-z-5,8,11,14-eicosatetraenoic acid, 5z,8z,11z,14z-icosa-5,8,11,14-tetraenoic acid, cis-5,8,11,14-eicosatetraenoic acid, 5,8,11,14-eicosatetraenoic acid, all-z, all-cis-5,8,11,14-eicosatetraenoic acid, unii-27yg812j1i, 5z,8z,11z,14z-eicosatetraenoic acid |
| InChI Key | YZXBAPSDXZZRGB-DOFZRALJSA-N |
| Molecular Weight (g/mol) | 304.474 |
| Formula Weight | 304.5 |
| Physical Form | Liquid |
| Chemical Name or Material | Arachidonic acid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?