missing translation for 'onlineSavingsMsg'
Learn More

Apigenin, MP Biomedicals

Product Code. 11422095 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422095 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11422095 Supplier MP Biomedicals Supplier No. 0520367180
  • ONLINE EXCLUSIVE PRICE
    190.00 EUR
    180.60 EUR / 100mg
    Save 9.40 EUR
    4% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 520-36-5
Molecular Formula C15H10O5
MDL Number MFCD00006831
Quantity 100 mg
Synonym apigenin, 5,7-dihydroxy-2-4-hydroxyphenyl-4h-chromen-4-one, chamomile, spigenin, versulin, apigenol, 4',5,7-trihydroxyflavone, apigenine, c.i. natural yellow 1, 5,7,4'-trihydroxyflavone
InChI Key KZNIFHPLKGYRTM-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C=C2O1
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
Molecular Weight (g/mol) 270.24
PubChem CID 5280443
ChEBI CHEBI:18388
Formula Weight 270.24
Chemical Name or Material Apigenin
Show More Show Less
compliance-icons
Product Identifier
  • APIGENIN, 100 MG
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4',5,7-Trihydroxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.