missing translation for 'onlineSavingsMsg'
Learn More

Aphidicolin, >99%, MP Biomedicals™

Product Code. 11450062 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450062 1 mg 1mg
11460062 5 mg 5mg
11470062 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450062 Supplier MP Biomedicals Supplier No. 0215988301

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Aphidicolin from Nigrospora oryzae ≥98%, Free Flowing Powder

  • It is an antibiotic, a potent antiviral and antimitotic agent and DNA polymerase inhibitor.
  • Inhibits the early S-phase of the cell cycle. DNA polymerase α and δ reversible inhibitor.
  • Potentiates apoptosis induced by arabinosyl nucleosides in leukemia cell lines.
  • Also induces apoptosis in HeLa S3 cells, but inhibits vincristine-induced apoptosis in the p53-negative human prostate cancer cell line PC-3.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38966-21-1
Molecular Formula C20H34O4
InChI Key NOFOAYPPHIUXJR-APNQCZIXSA-N
PubChem CID 457964
ChEBI CHEBI:2766
SMILES CC12CCC(C(C1CCC3C24CCC(C(C3)C4)(CO)O)(C)CO)O

Specifications

Color White
Quantity 1 mg
Synonym aphidicolin, unii-192tj6pp19, ccris 1783, chembl29711, 3r,4r,4ar,6as,8r,9r,11as,11bs-4,9-bis hydroxymethyl-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta a naphthalene-3,9-diol, 9,15-cyclo-c,18-dinor-14,15-secoandrostane-4,17-dimethanol, 3,17-dihydroxy-4-methyl-, 3alpha,4alpha,5alpha,17alpha, +-aphidicolin, 8,11a-methano-11ah-cyclohepta a naphthalene-4,9-dimethanol, tetradecahydro-3,9-dihydroxy-4,11b-dimethyl-, 3r-3-alpha,4-alpha,4a-alpha,6a-beta,8-beta,9-beta,11a-beta,11b-beta, +/--aphidicolin, aphidicolin, +/-
Molecular Weight (g/mol) 338.488
Formula Weight 338.5
Physical Form Powder
Chemical Name or Material Aphidicolin, For TLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.