Learn More
Apatinib-d8 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Apatinib-d8 Hydrochloride |
| CAS | 2468771-44-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24H16D8ClN5O |
| Quantity | 50 mg |
| Synonym | N-[4-(1-Cyanocyclopentyl-d8)phenyl]-2-[(4-pyridinylmethyl)amino]-3-pyridinecarboxamide, YN968D1-d8 Hydrochloride |
| InChI Formula | InChI=1S/C24H23N5O.ClH/c25-17-24(11-1-2-12-24)19-5-7-20(8-6-19)29-23(30)21-4-3-13-27-22(21)28-16-18-9-14-26-15-10-18;/h3-10,13-15H,1-2,11-12,16H2,(H,27,28)(H,29,30);1H/i1D2,2D2,11D2,12D2; |
| SMILES | O=C(C1=CC=CN=C1NCC2=CC=NC=C2)NC3=CC=C(C4(C#N)C([2H])([2H])C([2H])([2H])C([2H])([2H])C4([2H])[2H])C=C3.Cl |
| IUPAC Name | N-[4-(1-cyano-2,2,3,3,4,4,5,5-octadeuteriocyclopentyl)phenyl]-2-(pyridin-4-ylmethylamino)pyridine-3-carboxamide;hydrochloride |
| Molecular Weight (g/mol) | 441.98 |
| Show More |