Learn More
Apalutamide, MedChemExpress
Product Code. 16436978
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16436978 10 mg 10mg
16403037 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16436978 Supplier Medchem Express Supplier No. HY16060
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
MedChemExpress Apalutamide (ARN-509) is a potent and competitive androgen receptor (AR) antagonist, binding AR with an IC50 of 16 nM.
Apalutamide (ARN-509) is a potent and competitive androgen receptor (AR) antagonist, binding AR with an IC50 of 16 nM.
Specifications
Chemical Name or Material Apalutamide
CAS 956104-40-8
Color White
Purity Grade Notes Research
Molecular Formula C21H15F4N5O2S
Quantity 10 mg
Synonym ARN-509
Solubility Information DMSO : 50 mg/mL (104.73 mM; Need ultrasonic)
SMILES O=C(NC)C1=CC=C(N(C(N(C2=CC(C(F)(F)F)=C(C#N)N=C2)C3=O)=S)C43CCC4)C=C1F
Molecular Weight (g/mol) 477.43
Formula Weight 477.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death