Learn More
Antimony(V) Chloride, Sigma-Aldrich™
Product Code. 15849530
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15849530 100 mL 100mL
15839530 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15849530 Supplier Sigma Aldrich Supplier No. 8078370100
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7647-18-9
Grade Research
Molecular Weight (g/mol) 299.03
Specifications
Melting Point 3°C to 4°C
Quantity 100 mL
Density 2.35 g/cm3 at 20°C
MDL Number MFCD00011213
Synonym Antimony(V) chloride, Antimony pentachloride, Antimony(V) chloride
Chemical Name or Material Antimony(V) Chloride